Learn More
Biomatik Corporation Human Solute Carrier Family 39, Member 4 (SLC39A4) ELISA Kit
Supplier:  Biomatik Corporation EKU11436
Encompass_Preferred
Human Solute Carrier Family 39, Member 4 (SLC39A4) ELISA Kit for detecting SLC39A4 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.056ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q6P5W5. Gene ID: 55630. Target Synonyms: ZIP4; AEZ; Acrodermatitis Enteropathica,Zinc-Deficiency Type; Zinc Transporter Protein 4; Zinc transporter ZIP4; Zrt-and Irt-like protein 4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7148