Learn More
ACROBiosystems Human Sortilin / SORT1, His Tag (SON-H52H5) is expressed from human 293 cells (HEK293). It contains AA Ser 78 - Asn 755 (Accession # Q99523-1).
Catalog No. 502230808
Encompass_Preferred
The Sortilin is a sorting receptor in the Golgi compartment and as a clearance receptor on the cell surface. Required for protein transport from the Golgi apparatus to the lysosomes by a pathway that is independent of the mannose-6-phosphate receptor (M6PR). Lysosomal proteins bind specifically to the receptor in the Golgi apparatus and the resulting receptor-ligand complex is transported to an acidic prelysosomal compartment where the low pH mediates the dissociation of the complex. The receptor is then recycled back to the Golgi for another round of trafficking through its binding to the retromer.
This item is not returnable. View return policy
Catalog No. 50-223-0808 Supplier ACROBiosystems Supplier No. SONH52H5100UG
The Sortilin is a sorting receptor in the Golgi compartment and as a clearance receptor on the cell surface. Required for protein transport from the Golgi apparatus to the lysosomes by a pathway that is independent of the mannose-6-phosphate receptor (M6PR). Lysosomal proteins bind specifically to the receptor in the Golgi apparatus and the resulting receptor-ligand complex is transported to an acidic prelysosomal compartment where the low pH mediates the dissociation of the complex. The receptor is then recycled back to the Golgi for another round of trafficking through its binding to the retromer.