Learn More
Biomatik Corporation Human Sorting Nexin 13 (SNX13) ELISA Kit
Supplier:  Biomatik Corporation EKU07418
Encompass_Preferred
Human Sorting Nexin 13 (SNX13) ELISA Kit for detecting SNX13 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.128ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9Y5W8. Gene ID: 23161. Target Synonyms: RGS-PX1; RGS domain- and PHOX domain-containing protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6370