Learn More
Biomatik Corporation Human Sorting Nexin 17 (SNX17) ELISA Kit
Supplier:  Biomatik Corporation EKU07419
Encompass_Preferred
Human Sorting Nexin 17 (SNX17) ELISA Kit for detecting SNX17 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.059ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q15036. Gene ID: 9784. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7685