Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human SOX17 ELISA Kit

Catalog No. EH434RBX10
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH434RBX10 10 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EH434RBX10 Supplier Invitrogen™ Supplier No. EH434RBX10
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

ELISA

Human SOX17 quantitates human SOX17 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human SOX17.

SOX17 (transcription factor SOX-17) belongs to the SRY-related HMB box family of transcription factors. It acts as a transcription regulator that binds target promoter DNA and bends the DNA. It binds to the sequences 5'-AACAAT-'3 or 5'-AACAAAG-3'. SOX17 modulates transcriptional regulation via WNT3A and inhibits Wnt signaling. It promotes degradation of activated CTNNB1. SOX17 plays an essential role in the regulation of embryonic development and is required for normal looping of the embryonic hear tube and for normal development of the definitive gut endoderm. Mutations in the gene can result in vesicoureteral reflux.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9H6I2
Assay Range 0.03-25 ng/mL
Assay Sensitivity 0.03 ng/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Human
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias HMG transcription factor SOX17; HMG-box transcription factor Sox17; Sox17; Sox-17; SRY (sex determining region Y)-box 17; SRY box 17; SRY-box 17; SRY-box containing gene 17; SRY-box transcription factor 17; SRY-related HMG-box transcription factor SOX17; transcription factor SOX-17; transcription factor SOX-17; LOW QUALITY PROTEIN: transcription factor SOX-17; VUR3HMG transcription factor SOX17; HMG-box transcription factor Sox17; Sox17; Sox-17; SRY (sex determining region Y)-box 17; SRY box 17; SRY-box 17; SRY-box containing gene 17; SRY-box transcription factor 17; SRY-related HMG-box transcription factor SOX17; transcription factor SOX-17; transcription factor SOX-17; LOW QUALITY PROTEIN: transcription factor SOX-17; VUR3;
Gene ID (Entrez) 64321
Gene Symbol SOX17
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantity 10 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Target SOX17
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.