Learn More
Sino Biological Recombinant Human SOX2 Protein (His Tag)
Supplier:  Sino Biological 13890H07E1
Encompass_Preferred
A DNA sequence encoding the Human SOX2 (P48431) (Met1-Met317) was expressed, with a polyhistidine tag at the N-terminus.
Catalog No. 50-255-4171