Learn More
Raybiotech Inc Human SP-D ELISA Kit
Supplier:  Raybiotech Inc ELHSPD1
Encompass_Preferred
For Cell Culture Supernatants, Plasma, and Serum samples. Sensitivity: 0.12 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: The antibody pair provided in this kit recognizes Human Surfactant Pulmonary Associated Protein D (SFTPD)
Catalog No. 50-194-9747