Learn More
BIOHIPPO Human SP-D Microsample
SDP
Catalog No. 5000315772
Encompass
Human SP-D Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of SP-D in serum plasma tissue homogenates cell lysates lung lavage fluid cell culture supernates and other biological fluids It provides a detection range of 1 25-80 ng/mL with a sensitivity of 0 51 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human SP-D Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human SP-D Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human SP-D biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15772 Supplier BIOHIPPO Supplier No. BHE15210683
Human SP-D Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of SP-D in serum plasma tissue homogenates cell lysates lung lavage fluid cell culture supernates and other biological fluids It provides a detection range of 1 25-80 ng/mL with a sensitivity of 0 51 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human SP-D Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human SP-D Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human SP-D biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color