Learn More
Raybiotech Inc Human Sp1 Activity Assay Kit, 1 Plate Kit
Catalog No. NC3802104
Encompass_Preferred
RayBio® Human Sp1 Transcription Factor Activity Assay. This assay uses a dsDNA coated plate with canonical Sp1 binding sequences to semi-quantitatively detect active Sp1 in lysates or nuclear extracts. Dry ice shipment (additional fee).
Catalog No. NC3802104 Supplier Raybiotech Inc Supplier No. TFEHSP11
RayBio® Human Sp1 Transcription Factor Activity Assay. This assay uses a dsDNA coated plate with canonical Sp1 binding sequences to semi-quantitatively detect active Sp1 in lysates or nuclear extracts. Dry ice shipment (additional fee).