Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human SPANXC (aa 33-58) Control Fragment Recombinant Protein

Catalog No. RP99048
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP99048 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP99048 Supplier Invitrogen™ Supplier No. RP99048
  • $283.00
    $212.65 / Each of 1

    Save $70.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (42%), Rat (42%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-83819. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Temporally regulated transcription and translation of several testis-specific genes is required to initiate the series of molecular and morphological changes in the male germ cell lineage necessary for the formation of mature spermatozoa. This gene is a member of the SPANX family, which is located in a gene cluster on chromosome X. The SPANX genes encode differentially expressed testis-specific proteins that localize to various subcellular compartments. This particular gene encodes a protein that localizes to the nucleus and is expressed in highly metastatic cell lines, making the protein a potential diagnostic and prognostic marker. The protein belongs to a family of cancer/testis antigens and represents a potential target for cancer immunotherapy.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9NY87
Concentration ≥5.0 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 64663
Name Human SPANXC (aa 33-58) Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias C; cancer/testis antigen 11.3; cancer/testis antigen family 11, member 3; cancer/testis-associated protein CTp11; cancer/testis-associated protein of 11 kD; cancer-testis-associated protein CTp11; CT11.3; CTp11; nuclear-associated protein SPAN-Xc; SPANX family member C; SPANX family, member C; SPANXC; SPANX-C; SPAN-Xc protein; Sperm protein associated with the nucleus on the X chromosome C; sperm protein associated with the nucleus, X chromosome, family member C
Common Name SPANXC
Gene Symbol SPANXC
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence SDPQPAPKKLKTSESSTILVVRYRRN
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.