Learn More
Sino Biological Anti-SPARC Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10929T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human SPARC (rh SPARC; Catalog#10929-H08H; NP_003109.1; Met1-Ile303). SPARC specific IgG was purified by Human SPARC affinity chromatography.
Catalog No. 50-255-5065