Learn More
Sino Biological Anti-SPARC Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 200185T32100
Encompass_Preferred
Produced in rabbits immunized with E. coli-derived Human SPARC fragment, and purified by antigen affinity chromatography.
Catalog No. 50-241-1389