Learn More
American Research Products Inc Human SPARC related modular ca
Catalog No. 5000327170
Encompass
Human SPARC-related modular calcium-binding protein 2 (SMOC2) ELISA kit from CUSABIO Detection Range 0 156 ng/mL-10 ng/mL Sensitivity 0 039 ng/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich
Catalog No. 50-003-27170 Supplier American Research Products Inc Supplier No. CSBEL021843HU
Human SPARC-related modular calcium-binding protein 2 (SMOC2) ELISA kit from CUSABIO Detection Range 0 156 ng/mL-10 ng/mL Sensitivity 0 039 ng/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich