Learn More
Sino Biological Anti-SPARCL1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10046RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human SPARCL1 / MAST9 (rhSPARCL1; Catalog#10046-H08H; NP_004675.3; Met 1-Phe 664) . SPARCL1 specific IgG was purified by human SPARCL1 affinity chromatography.
Catalog No. 50-241-2355