Learn More
Biomatik Corporation Human S-Phase Kinase Associated Protein 1 (SKP1) ELISA Kit
Supplier:  Biomatik Corporation EKU07429
Encompass_Preferred
Human S-Phase Kinase Associated Protein 1 (SKP1) ELISA Kit for detecting SKP1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.062ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P63208. Gene ID: 6500. Target Synonyms: SKP1A; EMC19; OCP-II; OCP2; TCEB1L; p19A; Cyclin-A/CDK2-associated p19; RNA polymerase II elongation factor-like protein; Transcription elongation factor B polypeptide 1-like. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7034