Learn More
Biomatik Corporation Human S-Phase Kinase Associated Protein 2 (SKP2) ELISA Kit
Supplier:  Biomatik Corporation EKU07430
Encompass_Preferred
Human S-Phase Kinase Associated Protein 2 (SKP2) ELISA Kit for detecting SKP2 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.054ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q13309. Gene ID: 6502. Target Synonyms: FBL1; FBXL1; FLB1; p45; Cyclin-A/CDK2-associated protein p45; F-box/LRR-repeat protein 1; p45skp2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6170