Learn More
ACROBiosystems Unconjugated,Anti-SARS-CoV-2 Omicron Antibody-3A7C12,Human,Human IgG1 | Human Kappa,HEK is a chimeric monoclonal antibody recombinantly expressed from HEK293 cells, which combines the variable region of a mouse monoclonal antibody with huma
Catalog No. 502253559
Encompass_Preferred
It's been reported that Coronavirus can infect the human respiratory epithelial cells through interaction with the human ACE2 receptor. The spike protein is a large type I transmembrane protein containing two subunits, S1 and S2. S1 mainly contains a receptor binding domain (RBD), which is responsible for recognizing the cell surface receptor. S2 contains basic elements needed for the membrane fusion.The S protein plays key parts in the induction of neutralizing-antibody and T-cell responses, as well as protective immunity.
This item is not returnable. View return policy
Catalog No. 50-225-3559 Supplier ACROBiosystems Supplier No. SPDM415100UG
It's been reported that Coronavirus can infect the human respiratory epithelial cells through interaction with the human ACE2 receptor. The spike protein is a large type I transmembrane protein containing two subunits, S1 and S2. S1 mainly contains a receptor binding domain (RBD), which is responsible for recognizing the cell surface receptor. S2 contains basic elements needed for the membrane fusion.The S protein plays key parts in the induction of neutralizing-antibody and T-cell responses, as well as protective immunity.