Learn More
Brainxell Inc Spinal Motor Neurons (Human iPSC-Derived, Male Line, 1M)
SDP
Catalog No. NC2259381
Encompass
Human spinal motor neurons derived from iPSC line WC-30, male. Contents: 1 vial of ≥1x10^6 viable cryopreserved cells. Marker Expression: Spinal Motor Neurons (BX-0100) have high neuronal purity (>80%) and consist of greater than 70% motor neurons. Labeling with the pan-neuronal marker MAP2 (green) and the motor neuron-specific marker FOXP1 (red) highlights the purity of this neuronal product. Function: Spinal Motor Neurons exhibit pronounced electrophysiological activity after two weeks in culture, as demonstrated by multi-electrode array (MEA) recordings. Related diseases: Amyotrophic Lateral Sclerosis (ALS), Spinal muscular atrophy (SMA), & Spinal Cord Injury. Primary neurotransmitter released: Acetylcholine
Catalog No. NC2259381 Supplier Brainxell Inc Supplier No. BX0100301M
Human spinal motor neurons derived from iPSC line WC-30, male. Contents: 1 vial of ≥1x10^6 viable cryopreserved cells. Marker Expression: Spinal Motor Neurons (BX-0100) have high neuronal purity (>80%) and consist of greater than 70% motor neurons. Labeling with the pan-neuronal marker MAP2 (green) and the motor neuron-specific marker FOXP1 (red) highlights the purity of this neuronal product. Function: Spinal Motor Neurons exhibit pronounced electrophysiological activity after two weeks in culture, as demonstrated by multi-electrode array (MEA) recordings. Related diseases: Amyotrophic Lateral Sclerosis (ALS), Spinal muscular atrophy (SMA), & Spinal Cord Injury. Primary neurotransmitter released: Acetylcholine