Learn More
Cusabio Technology LLC HUMAN SPINK1 ELISA KIT
Catalog No. 5000049684
Encompass_Preferred
This ELISA kit is designed for the quantitative detection of Human Pancreatic Secretory Trypsin Inhibitor (SPINK1) in serum plasma and tissue homogenates using a sandwich assay format It offers a broad detection range from 78 pg/ml to 5000 pg/ml with a sensitivity limit of 19 5 pg/ml ensuring precise measurement across various sample concentrations The assay requires a short incubation period of 1-5 hours and utilizes a sample volume of 50-100 l per test Optical density is measured at 450 nm for result interpretation providing reliable and reproducible data for research applications requiring SPINK1 level analysis in biological samples
Catalog No. 50-000-49684 Supplier Cusabio Technology LLC Supplier No. CSBEL022575HU
This ELISA kit is designed for the quantitative detection of Human Pancreatic Secretory Trypsin Inhibitor (SPINK1) in serum plasma and tissue homogenates using a sandwich assay format It offers a broad detection range from 78 pg/ml to 5000 pg/ml with a sensitivity limit of 19 5 pg/ml ensuring precise measurement across various sample concentrations The assay requires a short incubation period of 1-5 hours and utilizes a sample volume of 50-100 l per test Optical density is measured at 450 nm for result interpretation providing reliable and reproducible data for research applications requiring SPINK1 level analysis in biological samples