Learn More
Sino Biological Anti-HAI-1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 11742RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human HAI-1 / SPINT1 (rh HAI-1 / SPINT1; Catalog#11742-H08H; O43278-2; Met 1-Val 433). HAI-1 / SPINT1 specific IgG was purified by Human HAI-1 / SPINT1 affinity chromatography.
Catalog No. 50-255-4585