Learn More
Sino Biological Anti-HAI-1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 201756T08100
Encompass_Preferred
Produced in rabbits immunized with E. coli-derived Human HAI-1 / SPINT1 fragment, and purified by antigen affinity chromatography.
Catalog No. 50-255-3936