Learn More
Sino Biological Human SPINT2 Protein 50ug (Sino Biological)
Supplier:  Sino Biological 10324H02H50
Encompass_Preferred
A DNA sequence encoding the human SPINT2 (NP_066925.1) (Met1-Lys197) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-237-2555