Learn More
Zymo Research Corporation Human Spleen DNA (5 ug)
SDP
Catalog No. 502037114
Encompass_Preferred
This set is an ideal control for detection and/or quantification methods against 5- methylcytosine (5mC) and 5-hydroxymethylcytosine (5hmC) as both modified cytosines are present at physiologically relevant levels and loci.
Catalog No. 50-203-7114 Supplier Zymo Research Corporation Supplier No. D50182
This set is an ideal control for detection and/or quantification methods against 5- methylcytosine (5mC) and 5-hydroxymethylcytosine (5hmC) as both modified cytosines are present at physiologically relevant levels and loci.