Learn More
BIORTUS USA INC Human SPR Protein, 100UG
Catalog No. NC2953348
Encompass_Preferred
Gene Name: SPR. Uniprot: P35270. Construct: 6His-TEV-SPR(M1-K261 end). Expression Host: E. coli. Purity: 90%.
Catalog No. NC2953348 Supplier BIORTUS USA INC Supplier No. BP0580701A
Gene Name: SPR. Uniprot: P35270. Construct: 6His-TEV-SPR(M1-K261 end). Expression Host: E. coli. Purity: 90%.