Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human SREC-I/SCARF1 ELISA Kit

Catalog No. EH437RBX10
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH437RBX10 10 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EH437RBX10 Supplier Invitrogen™ Supplier No. EH437RBX10
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

ELISA

Human SREC-I/SCARF1 quantitates human SREC-I/SCARF1 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human SREC-I/SCARF1.

The protein encoded by this gene is a scavenger receptor that is expressed in endothelial cells and mediates the binding and degradation of acetylated low density lipoprotein (Ac-LDL). Transcription factors ZNF444/EZF-2 and SP1 are found to be responsible for the regulation of this gene. Five alternatively spliced transcript variants encoding distinct isoforms have been described.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q14162
Assay Range 0.08-20 ng/mL
Assay Sensitivity 0.08 ng/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Human
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias AA986099; acetyl LDL receptor; endothelial cells scavenger receptor; KIAA0149; MGC47738; mKIAA0149; SCARF1; Scavenger receptor class F member 1; scavenger receptor class F, member 1; scavenger receptor expressed by endothelial cells 1; Scavenger receptor expressed by endothelial cells 1 protein; SREC; SREC1; SREC-IAA986099; acetyl LDL receptor; endothelial cells scavenger receptor; KIAA0149; MGC47738; mKIAA0149; SCARF1; Scavenger receptor class F member 1; scavenger receptor class F, member 1; scavenger receptor expressed by endothelial cells 1; Scavenger receptor expressed by endothelial cells 1 protein; SREC; SREC1; SREC-I;
Gene ID (Entrez) 8578
Gene Symbol Scarf1
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantity 10 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Target SCARF1
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.