Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human SSH2 (aa 1008-1101) Control Fragment Recombinant Protein

Catalog No. RP103721
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP103721 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP103721 Supplier Invitrogen™ Supplier No. RP103721
  • $283.00
    $212.65 / Each of 1

    Save $70.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (65%), Rat (65%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-63402. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

SSH2 (slingshot homolog 2) is a 1,423 amino acid protein that localizes to both the cytoplasm and the cytoskeleton and is a human homolog of the Drosophila slingshot (ssh) protein. Containing one tyrosine-protein phosphatase domain, SSH2 functions as a protein phosphatase that regulates Actin filament dynamics via the dephosphorylation of target proteins, such as Cofilin, which mediate Actin filament assembly and disassembly. Four isoforms of SSH2 exist due to alternative splicing events. The gene encoding SSH2 maps to human chromosome 17, which comprises over 2.5% of the human genome and encodes over 1,200 genes. Two key tumor suppressor genes are associated with chromosome 17, namely, p53 and BRCA1. Tumor suppressor p53 is necessary for maintenance of cellular genetic integrity by moderating cell fate through DNA repair versus cell death. Malfunction or loss of p53 expression is associated with malignant cell growth and Li-Fraumeni syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q76I76
Concentration ≥5.0 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 85464
Name Human SSH2 (aa 1008-1101) Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias hSSH-2L; KIAA1725; mSSH-2L; Protein phosphatase Slingshot homolog 2; slingshot 2L; slingshot homolog 2; slingshot homolog 2 (Drosophila); slingshot protein phosphatase 2; slingshot-like 2; Ssh2; SSH-2; Ssh2l; SSH-2L; SSH-like protein 2
Common Name SSH2
Gene Symbol SSH2
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence RVEIIEYTHIVTSPNHTGPGSEIATSEKSGEQGLRKVNMEKSVTVLCTLDENLNRTLDPNQVSLHPQVLPLPHSSSPEHNRPTDHPTSILSSPE
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.