Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human ST8SIA2 ELISA Kit

Catalog No. EH440RBX5
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH440RBX5 5 x 96 Tests
EH440RBX10 10 x 96 Tests
EH440RB 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. EH440RBX5 Supplier Invitrogen™ Supplier No. EH440RBX5
Get Up to $3,000 in Extra Life Sciences Consumables with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ELISA

Human ST8SIA2 quantitates human ST8SIA2 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human ST8SIA2.

The protein encoded by this gene is a type II membrane protein that is thought to catalyze the transfer of sialic acid from CMP-sialic acid to N-linked oligosaccharides and glycoproteins. The encoded protein may be found in the Golgi apparatus and may be involved in the production of polysialic acid, a modulator of the adhesive properties of neural cell adhesion molecule (NCAM1). This protein is a member of glycosyltransferase family 29.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q92186
Assay Range 0.62-150 ng/mL
Assay Sensitivity 0.62 ng/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Human
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias AI323367; alpha-2,8-sialyltransferase; alpha-2,8-sialyltransferase 8B; alpha-2,8-sialyltransferase 8B 1; cb394; HsT19690; id:ibd5161; MGC116854; MGC116857; N-glycan alpha 2,8-sialyltransferase; polysialic acid synthase; sialyltransferase 8; sialyltransferase 8 (alpha-2 8-sialytransferase) B; sialyltransferase 8 (alpha-2, 8-sialyltransferase) B; sialyltransferase 8 (alpha-2, 8-sialytransferase) B; sialyltransferase 8B; Sialyltransferase St8Sia II; sialyltransferase X; sialytransferase St8Sia II; SIAT 8B; siat8; SIAT8B; SIAT8-B; ST8 alpha-N-acetylneuraminate alpha-2,8-sialyltransferase 2; ST8 alpha-N-acetyl-neuraminide alpha-2,8-sialyltransferase 2; ST8Sia II; ST8SIA2; st8sia2 protein; ST8SiaII; ST8SIA-II; STXAI323367; alpha-2,8-sialyltransferase; alpha-2,8-sialyltransferase 8B; alpha-2,8-sialyltransferase 8B 1; cb394; HsT19690; id:ibd5161; MGC116854; MGC116857; N-glycan alpha 2,8-sialyltransferase; polysialic acid synthase; sialyltransferase 8; sialyltransferase 8 (alpha-2 8-sialytransferase) B; sialyltransferase 8 (alpha-2, 8-sialyltransferase) B; sialyltransferase 8 (alpha-2, 8-sialytransferase) B; sialyltransferase 8B; Sialyltransferase St8Sia II; sialyltransferase X; sialytransferase St8Sia II; SIAT 8B; siat8; SIAT8B; SIAT8-B; ST8 alpha-N-acetylneuraminate alpha-2,8-sialyltransferase 2; ST8 alpha-N-acetyl-neuraminide alpha-2,8-sialyltransferase 2; ST8Sia II; ST8SIA2; st8sia2 protein; ST8SiaII; ST8SIA-II; STX;
Gene ID (Entrez) 8128
Gene Symbol ST8SIA2
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Target ST8SIA2
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.