Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ST8SIA5 Partial ORF (NP_037437, 33 a.a. - 132 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. p-7100185 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-963-412 10 μg
89-963-413 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-963-412 Supplier Abnova™ Supplier No. H00029906Q01S

Used for AP, Array, ELISA, WB-Re

The protein encoded by this gene is a type II membrane protein that may be present in the Golgi apparatus. The encoded protein, which is a member of glycosyltransferase family 29, may be involved in the synthesis of gangliosides GD1c, GT1a, GQ1b, and GT3 from GD1a, GT1b, GM1b, and GD3, respectively. [provided by RefSeq]

Sequence: QQILYGRNYIKRYFEFYEGPFEYNSTRCLELRHEILEVKVLSMVKQSELFDRWKSLQMCKWAMNISEANQFKSTLSRCCNAPAFLFTTQKNTPLGTKLKY

Specifications

Accession Number NP_037437
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 29906
Molecular Weight (g/mol) 36.74kDa
Name ST8SIA5 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen QQILYGRNYIKRYFEFYEGPFEYNSTRCLELRHEILEVKVLSMVKQSELFDRWKSLQMCKWAMNISEANQFKSTLSRCCNAPAFLFTTQKNTPLGTKLKY
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias MGC119670/MGC119671/SIAT8E/ST8SiaV
Common Name ST8SIA5
Gene Symbol ST8SIA5
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.