Learn More
Sino Biological Anti-STAT3 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 109668T32100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human STAT3, and purified by antigen affinity chromatography.
Catalog No. 50-255-5020