Learn More
Sino Biological Recombinant Human STAT3 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 10034H07E100
Encompass_Preferred
A DNA sequence encoding the human STAT3 (P40763-1) (Gly127-Arg688) was expressed, with a polyhistidine tag at the N-terminus.
Catalog No. 50-237-2586