Learn More
Sino Biological Human STAT6 MAb 100ul (Sino Biological)
Supplier:  Sino Biological 13190MM02100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human STAT6 (rh STAT6; Catalog#13190-H08B; P42226-1; Met1-Trp847). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-3124