Learn More
Sino Biological Anti-STC1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 15411RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human STC1 (rh STC1; Catalog#15411-H08H; NP_003146.1; Met1-Ala247). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-4105