Learn More
BIOHIPPO Human STC2 Microsample
SDP
Catalog No. 5000315883
Encompass
Human STC2 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of STC2 in serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 31 25-2000 pg/mL with a sensitivity of 12 6 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human STC2 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human STC2 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human STC2 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15883 Supplier BIOHIPPO Supplier No. BHE15211675
Human STC2 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of STC2 in serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 31 25-2000 pg/mL with a sensitivity of 12 6 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human STC2 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human STC2 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human STC2 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color