Learn More
Sino Biological Anti-STC2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 13653T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human STC2 (rh STC2; Catalog#13653-H08H; O76061; Met1-Arg302). STC2 specific IgG was purified by Human STC2 affinity chromatography.
Catalog No. 50-255-4196