Learn More
Sino Biological Anti-STC2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 13653RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human STC2 (rh STC2; Catalog#13653-H08H; O76061; Met1-Arg302). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-4197