Learn More
ACROBiosystems Human STEAP1 Full Length Protein, Twin-Strep,His Tag (Detergent)
Catalog No. 502718852
Encompass_Preferred
Six-transmembrane epithelial antigen of the prostate 1 (STEAP1) is an integral membrane protein that is highly up-regulated on the cell surface of several human cancers, making it a promising therapeutic target to manage these diseases. It shares sequence homology with three enzymes (STEAP2-STEAP4) that catalyze the NADPH-dependent reduction of iron(III). Taking into account its high specificity and overexpression in human cancers, STEAP1 is nowadays a promising candidate to be imposed as a therapeutic target.
Catalog No. 50-271-8852 Supplier ACROBiosystems Supplier No. ST1H5283100UG
Six-transmembrane epithelial antigen of the prostate 1 (STEAP1) is an integral membrane protein that is highly up-regulated on the cell surface of several human cancers, making it a promising therapeutic target to manage these diseases. It shares sequence homology with three enzymes (STEAP2-STEAP4) that catalyze the NADPH-dependent reduction of iron(III). Taking into account its high specificity and overexpression in human cancers, STEAP1 is nowadays a promising candidate to be imposed as a therapeutic target.