Learn More
ACROBiosystems HEK293/Human STEAP1 Stable Cell Line
Catalog No. 502719746
Encompass_Preferred
Six-transmembrane epithelial antigen of the prostate 1 (STEAP1) is an integral membrane protein that is highly up-regulated on the cell surface of several human cancers, making it a promising therapeutic target to manage these diseases. It shares sequence homology with three enzymes (STEAP2-STEAP4) that catalyze the NADPH-dependent reduction of iron(III). Taking into account its high specificity and overexpression in human cancers, STEAP1 is nowadays a promising candidate to be imposed as a therapeutic target.
Catalog No. 50-271-9746 Supplier ACROBiosystems Supplier No. CHEKATP1542VIALS
 Restricted Item
Add to Cart
Add to Cart
Six-transmembrane epithelial antigen of the prostate 1 (STEAP1) is an integral membrane protein that is highly up-regulated on the cell surface of several human cancers, making it a promising therapeutic target to manage these diseases. It shares sequence homology with three enzymes (STEAP2-STEAP4) that catalyze the NADPH-dependent reduction of iron(III). Taking into account its high specificity and overexpression in human cancers, STEAP1 is nowadays a promising candidate to be imposed as a therapeutic target.