Learn More
Raybiotech Inc C-Series Human Stem Cell Array 1 Kit.
Supplier:  Raybiotech Inc AAHSC12
Encompass_Preferred
Semi-quantitative measurement of 15 human stem cell biomarkers. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. Proteins detected: Alpha-fetoprotein, BMPR-IA (ALK-3), ALK-6 (BMPR-IB), Brachyury (Protein T), CD38, E-Cadherin, GATA4, hCG beta, Nanog, Nestin, OCT-4, PDX-1 (IPF1), SOX17, SOX2, VEGFR2
Catalog No. 50-194-8649