Learn More
Raybiotech Inc G-Series Human Stem Cell Array 1 Kit.
Catalog No. 501948653
Encompass_Preferred
Semi-quantitative measurement of 15 human stem cell biomarkers. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. Proteins detected: Alpha-fetoprotein, OCT-4, BMPR-IA (ALK-3), ALK-6 (BMPR-IB), Brachyury (Protein T), CD38, E-Cadherin, GATA4, hCG beta, Nanog, Nestin, PDX-1 (IPF1), SOX2, SOX17, VEGFR3
Catalog No. 50-194-8653 Supplier Raybiotech Inc Supplier No. AAHSCG18
Semi-quantitative measurement of 15 human stem cell biomarkers. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. Proteins detected: Alpha-fetoprotein, OCT-4, BMPR-IA (ALK-3), ALK-6 (BMPR-IB), Brachyury (Protein T), CD38, E-Cadherin, GATA4, hCG beta, Nanog, Nestin, PDX-1 (IPF1), SOX2, SOX17, VEGFR3