Learn More
Biomatik Corporation Human Stem Cell Factor Receptor (SCFR) CLIA Kit
Supplier:  Biomatik Corporation EKU08836
Encompass_Preferred
Human Stem Cell Factor Receptor (SCFR) CLIA Kit for detecting SCFR in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 41.2-30,000pg/ml. Sensitivity: 15.3pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P10721. Gene ID: 3815. Target Synonyms: CD117; KIT; C-Kit; PBT; p145 c-Kit; C-Kit Receptor; Tyrosine-Protein Kinase; V-Kit Hardy-Zuckerman 4 Feline Sarcoma Viral Oncogene Homolog; Piebald Trait. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6861