Learn More
Lonza Walkersville Human Stem Cell Nucleofector Kit 1
Catalog No. NC0566431
Encompass
The Nucleofector 2b Device (or older generations I or II) works with cell type specific kits, each of them dedicated to an individual primary cell. The Human Stem Cell Nucleofector Kit 1 is one of our two kits suited for efficient transfection of human stem cells. Due to significant diversity between different human stem cell clones, we have developed two Nucleofector Kits (1 and 2) and an easy-to-use Nucleofector Starter Kit to determine which of the 2 kits is optimal for a specific clone.Benefits:High transfection efficiencies for primary cellsCells maintain functionality post transfectionApplications:Transfection of cell numbers in the range of 2 x 10⁵ to 2 x 10⁷ cells Low-throughput transfection in single cuvette formatContent:2.25 mL Nucleofector Solution for Human Stem Cells0.5 mL Supplement 1, 30 µg pmaxGFP Vector (0.5 μg/μl in 10 mM Tris pH 8.0)25 Aluminum cuvette (100 µL)25 Single use pipettes
Catalog No. NC0566431 Supplier Lonza Walkersville Supplier No. VPH5012
The Nucleofector 2b Device (or older generations I or II) works with cell type specific kits, each of them dedicated to an individual primary cell. The Human Stem Cell Nucleofector Kit 1 is one of our two kits suited for efficient transfection of human stem cells. Due to significant diversity between different human stem cell clones, we have developed two Nucleofector Kits (1 and 2) and an easy-to-use Nucleofector Starter Kit to determine which of the 2 kits is optimal for a specific clone.Benefits:High transfection efficiencies for primary cellsCells maintain functionality post transfectionApplications:Transfection of cell numbers in the range of 2 x 10⁵ to 2 x 10⁷ cells Low-throughput transfection in single cuvette formatContent:2.25 mL Nucleofector Solution for Human Stem Cells0.5 mL Supplement 1, 30 µg pmaxGFP Vector (0.5 μg/μl in 10 mM Tris pH 8.0)25 Aluminum cuvette (100 µL)25 Single use pipettes