Learn More
MilliporeSigma™ Human STEMCCA Cre-Excisable Constitutive (OKSM) Lentivirus
Catalog No. SCR5451 Shop All MilliporeSigma Products
Change view
Click to view available options
Human STEMCCA Cre-Excisable Constitutive (OKSM) Lentivirus
Catalog No.
SCR5451 Human STEMCCA Cre-Excisable Constitutive (OKSM) Lentivirus
1 options
Catalog No. SCR5451 Supplier MilliporeSigma™ Supplier No. SCR5451
For use with Human STEMCCA Cre-Excisable Constitutive Polycistronic (OKSM) Lentivirus Reprogramming Kit (SCR545).
  • Used with Human STEMCCA Cre-Excisable Constitutive Polycistronic (OKSM) Lentivirus Reprogramming Kit (SCR545)
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Lentivirus is stable for at least 6 months when stored at -80°C. After first thaw, place immediately on ice and store in working aliquots to avoid further freeze thaws. Avoid freeze thaws as this will result in a decrease in the virus titer. Polybrene transfection reagent is stable for at least 1 year when stored at -20°C.
Product Type Lentivirus Reprogramming Kit