Learn More
Biomatik Corporation Human Steroid 5 Alpha Reductase 1 (SRD5a1) ELISA Kit
Supplier:  Biomatik Corporation EKU11442
Encompass_Preferred
Human Steroid 5 Alpha Reductase 1 (SRD5a1) ELISA Kit for detecting SRD5a1 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.114ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P18405. Gene ID: 6715. Target Synonyms: 3-Oxo-5 Alpha-Steroid Delta 4-Dehydrogenase Alpha 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6474