Learn More
Sino Biological Anti-LOK Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 11724T52100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human LOK (rh LOK; Catalog#11724-H07E; NP_005981.3; Arg18-Glu317). LOK specific IgG was purified by Human LOK affinity chromatography.
Catalog No. 50-255-4602