Learn More
Sino Biological Human STK4 MAb 100ul (Sino Biological)
Supplier:  Sino Biological 11533MM04100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human STK4 / MST1 (rh STK4 / MST1; Catalog#11533-H07B; Q13043-1; Glu 2-Phe 487). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-1892