Learn More
Sino Biological Anti-MST1 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 11533MM0620
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human STK4 / MST1 (rh STK4 / MST1; Catalog#11533-H07B; Q13043-1; Glu2-Phe487). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-255-4701