Learn More
Sino Biological Human STXBP1 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 11751T20100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human Munc18-1 / STXBP1 (rh Munc18-1 / STXBP1; Catalog#11751-H20B; P61764-1; Met1-Ser594). Munc18-1 / STXBP1 specific IgG was purified by Human Munc18-1 / STXBP1 affinity chromatography.
Catalog No. 50-237-2036