Learn More
Biomatik Corporation Human Substance P (SP) CLIA Kit
Supplier:  Biomatik Corporation EKU08615
Encompass_Preferred
Human Substance P (SP) CLIA Kit for detecting SP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 7.81-2,000pg/ml. Sensitivity: 2.86pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Chemiluminescence. Assay Time: 2h. Accession Number: P20366. Gene ID: 6863. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5067