Learn More
Biomatik Corporation Human Succinate Dehydrogenase Complex Subunit B (SDHB) ELISA Kit
Supplier:  Biomatik Corporation EKU11810
Encompass_Preferred
Human Succinate Dehydrogenase Complex Subunit B (SDHB) ELISA Kit for detecting SDHB in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.121ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P21912. Gene ID: 6390. Target Synonyms: SDH1; SDH; Ip; Iron Sulfur; Iron-sulfur subunit of complex II; Succinate dehydrogenase [ubiquinone] iron-sulfur subunit, mitochondrial. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4511