Learn More
Sino Biological Anti-SUGCT Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 103670T08100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human SUGCT, and purified by antigen affinity chromatography.
Catalog No. 50-241-2015